Please login to your online account to display your discounted pricing

R&D Systems Rat Erythropoietin Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 24335
Molecular Weight 18.5 kDa
Name Erythropoietin
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1306RE010

 
r&d systems
RDS1306RE010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87

1306RE010CF

 
r&d systems
RDS1306RE010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 24335
Molecular Weight 18.5 kDa
Name Erythropoietin
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
Recombinant Yes
Source Rat

Source: Spodoptera frugiperda, Sf 21 (stably transfected)-derived, Ala27-Arg192
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 24335
Regulatory Status: RUO