missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat EGF Recombinant Protein

$285.48 - $285.48

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 25313
Molecular Weight 6.3 kDa
Name EGF
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
3214EG100 R&D Systems
RDS3214EG100
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $285.48
3214EG100CF R&D Systems
RDS3214EG100CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $285.48
Specifications

Description

Source: E. coli-derived, Asn974-Arg1026, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 6.3 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 25313
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
6.3 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
25313
EGF
100μg
Rat