missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat CXCL3/CINC-2 alpha Recombinant Protein

$376.73 - $376.73

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 171551
Molecular Weight 7.6 kDa
Name CXCL3/CINC-2 alpha
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
516CA025 R&D Systems
RDS516CA025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $376.73
516CA025CF R&D Systems
RDS516CA025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $376.73
Specifications

Description

Source: E. coli-derived, Arg33-Ser100
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 171551
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
7.6 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
171551
CXCL3/CINC-2 alpha
25μg
Rat