missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat CXCL2/GRO beta/MIP-2/CINC-3 Recombinant Protein

$298.51 - $298.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 114105
Molecular Weight 7.6 kDa
Name CXCL2/GRO beta/MIP-2/CINC-3
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
525C3025CF R&D Systems
RDS525C3025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $298.51
525C3025 R&D Systems
RDS525C3025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $298.51
Specifications

Description

Source: E. coli-derived, Ser32-Asn100
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 114105
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
7.6 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
114105
CXCL2/GRO beta/MIP-2/CINC-3
25μg
Rat