missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat CX3CL1/Fractalkine Chemokine Domain, aa 25-100 Recombinant Protein

$404.12 - $404.12

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 89808
Molecular Weight 8.8 kDa
Name CX3CL1/Fractalkine Chemokine Domain, aa 25-100
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
537FT025CF R&D Systems
RDS537FT025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $404.12
537FT025 R&D Systems
RDS537FT025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $404.12
Specifications

Description

Source: E. coli-derived, Gln25-Gly100
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 89808
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
8.8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
89808
CX3CL1/Fractalkine Chemokine Domain, aa 25-100
25μg
Rat