missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat CX3CL1/Fractalkine Chemokine Domain, aa 22-100 Recombinant Protein

$415.83 - $415.83

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 89808
Molecular Weight 9.0 kDa
Name CX3CL1/Fractalkine Chemokine Domain, aa 22-100
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
568FR025 R&D Systems
RDS568FR025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $415.83
568FR025CF R&D Systems
RDS568FR025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $415.83
Specifications

Description

Source: E. coli-derived, Leu22-Gly100
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 9.0 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 89808
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
9.0 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
89808
CX3CL1/Fractalkine Chemokine Domain, aa 22-100
25μg
Rat