missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat CNTF Recombinant Protein

$220.30 - $663.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 25707
Molecular Weight 22.8 kDa
Name CNTF
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
557NT010 R&D Systems
RDS557NT010
Lyophilized from a 0.2μm filtered solution in Tris, NaCl, TCEP, EDTA, CHAPS and Trehalose with BSA as a carrier protein. 10μg Each for $220.30
557NT010CF R&D Systems
RDS557NT010CF
Lyophilized from a 0.2μm filtered solution in Tris, NaCl, TCEP, EDTA, CHAPS and Trehalose. 10μg Each for $220.30
557NT050 R&D Systems
RDS557NT050
Lyophilized from a 0.2μm filtered solution in Tris, NaCl, TCEP, EDTA, CHAPS and Trehalose with BSA as a carrier protein. 50μg Each for $663.51
557NT050CF R&D Systems
RDS557NT050CF
Lyophilized from a 0.2μm filtered solution in Tris, NaCl, TCEP, EDTA, CHAPS and Trehalose. 50μg Each for $663.51
Specifications

Description

Source: E. coli-derived, Ala2-Met200
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 22.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Tris, NaCl, TCEP, EDTA, CHAPS and Trehalose.
Entrez Gene ID: 25707
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
22.8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
25707
CNTF
Yes