missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat CNTF R alpha Recombinant Protein

$454.80 - $454.80

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 313173
Molecular Weight 39.1 kDa
Name CNTF R alpha
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
558CR050 R&D Systems
RDS558CR050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $454.80
558CR050CF R&D Systems
RDS558CR050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $454.80
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Ala19-Pro346 & Gln23-Pro346
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 39.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 313173
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
39.1 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
313173
CNTF R alpha
50μg
Rat