Please login to your online account to display your discounted pricing

R&D Systems Rat CCL4/MIP-1 beta Recombinant Protein

$298.51 - $298.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 116637
Molecular Weight 7.8 kDa
Name CCL4/MIP-1 beta
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

6935MB025

 
r&d systems
RDS6935MB025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $298.51

6935MB025CF

 
r&d systems
RDS6935MB025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $298.51
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 116637
Molecular Weight 7.8 kDa
Name CCL4/MIP-1 beta
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Rat

Source: E. coli-derived, Ala24-Asn92
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 116637
Regulatory Status: RUO