missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat CCL3/MIP-1 alpha Recombinant Protein

$298.51 - $298.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 25542
Molecular Weight 7.8 kDa
Name CCL3/MIP-1 alpha
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
6625MA025 R&D Systems
RDS6625MA025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $298.51
6625MA025CF R&D Systems
RDS6625MA025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $298.51
Specifications

Description

Source: E. coli-derived, Ala24-Ala92
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 25542
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
7.8 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
25542
CCL3/MIP-1 alpha
25μg
Rat