missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat CCL27/CTACK Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 362505
Molecular Weight 10.9 kDa
Name CCL27/CTACK
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
4434CK025CF R&D Systems
RDS4434CK025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
4434CK025 R&D Systems
RDS4434CK025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
Specifications

Description

Source: E. coli-derived, Leu70-Lys164
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 10.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 362505
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
10.9 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
362505
CCL27/CTACK
25μg
Rat