missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat CCL20/MIP-3 alpha Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 29538
Molecular Weight 8.2 kDa
Name CCL20/MIP-3 alpha
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
540RM025CF R&D Systems
RDS540RM025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $428.87
540RM025 R&D Systems
RDS540RM025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $428.87
Specifications

Description

Source: E. coli-derived, Ala26-Met96
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.2 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 29538
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
8.2 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
29538
CCL20/MIP-3 alpha
25μg
Rat