Please login to your online account to display your discounted pricing

R&D Systems Rat CCL19/MIP-3 beta Recombinant Protein

$368.77 - $368.77

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 362506
Molecular Weight 9.1 kDa
Name CCL19/MIP-3 beta
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

7800M3025

 
r&d systems
RDS7800M3025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $368.77

7800M3025CF

 
r&d systems
RDS7800M3025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $368.77
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 362506
Molecular Weight 9.1 kDa
Name CCL19/MIP-3 beta
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Rat

Source: E. coli-derived, Gly26-Ser108
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 9.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 362506
Regulatory Status: RUO