missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat C-Reactive Protein/CRP Recombinant Protein

$311.55 - $311.55

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 25419
Molecular Weight 24 kDa (monomer)
Name C-Reactive Protein/CRP
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1744CR200CF R&D Systems
RDS1744CR200CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $311.55
1744CR200 R&D Systems
RDS1744CR200
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $311.55
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, His20-Ser230, with a C-terminal 6-His tag
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 24 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 25419
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
24 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
25419
C-Reactive Protein/CRP
200μg
Rat