missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat Agrin Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 25592
Molecular Weight 90 kDa
Name Agrin
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
550AG100 R&D Systems
RDS550AG100
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $416.79
550AG100CF R&D Systems
RDS550AG100CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $416.79
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Ala1153-Pro1959 (Pro1788-Ser1798 del), with an N-terminal Met and 6-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 90 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 25592
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
90 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
25592
Agrin
100μg
Rat