missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rabbit IL-4 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 100302454
Molecular Weight 14.2 kDa
Name IL-4
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
6939RB025 R&D Systems
RDS6939RB025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
6939RB025CF R&D Systems
RDS6939RB025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: E. coli-derived, Arg25-Ser147, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14.2 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 100302454
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
14.2 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
100302454
IL-4
25μg
Rabbit