missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rabbit IL-21 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 100344172
Molecular Weight 15.5 kDa
Name IL-21
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
7274RB025 R&D Systems
RDS7274RB025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
7274RB025CF R&D Systems
RDS7274RB025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: E. coli-derived, His136-Arg268, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 100344172
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
15.5 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
100344172
IL-21
25μg
Rabbit