missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rabbit IL-2 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 100302458
Molecular Weight 15.2 kDa
Name IL-2
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
6994R2020CF R&D Systems
RDS6994R2020CF
Lyophilized from a 0.2μm filtered solution in Sodium Acetate. Each for $428.87
6994R2020 R&D Systems
RDS6994R2020
Lyophilized from a 0.2μm filtered solution in Sodium Acetate with BSA as a carrier protein. Each for $428.87
Specifications

Description

Source: E. coli-derived, Ala21-Ser153, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.2 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Sodium Acetate.
Entrez Gene ID: 100302458
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
15.2 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
100302458
IL-2
20μg
Rabbit