missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rabbit IL-1 beta/IL-1F2 Recombinant Protein

$376.73 - $376.73

Specifications

Conjugate Unlabeled
Molecular Weight 17.5 kDa
Name IL-1 beta/IL-1F2
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
7464RB010 R&D Systems
RDS7464RB010
Lyophilized from a 0.2μm filtered solution in MES, NaCl, DTT and EDTA with BSA as a carrier protein. Each for $376.73
7464RB010CF R&D Systems
RDS7464RB010CF
Lyophilized from a 0.2μm filtered solution in MES, NaCl, DTT and EDTA. Each for $376.73
Specifications

Description

Source: E. coli-derived, Ala117-Ser268
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MES, NaCl, DTT and EDTA.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
IL-1 beta/IL-1F2
10μg
Rabbit
17.5 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes