Please login to your online account to display your discounted pricing

R&D Systems ProDots Recombinant Human SCF/c-kit Ligand


Pre-aliquoted balls of SCF protein for cell culture media

Manufacturer: r&d systems  PRD25550

 View more versions of this product

Catalog No. PRD25550

  • / Each
 

Description & Specifications

Specifications

Recombinant Yes
Purity >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Storage Requirements Stored at 4°C where they will remain stable for 6 months.
Quantity 250μg
Molecular Weight 18.6kDa
Name ProDots™ Recombinant Human SCF
Source E. coli-derived,Glu26-Ala189, with an N-terminal Met
Accession Number P21583.1