Please login to your online account to display your discounted pricing

R&D Systems ProDots Recombinant Human SCF/c-kit Ligand


Pre-aliquoted balls of SCF protein for cell culture media

Manufacturer: r&d systems  PRD25510

 View more versions of this product

Catalog No. PRD25510

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 18.6kDa
Purity >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
Recombinant Yes
Source Human
Storage Requirements Stored at 4°C where they will remain stable for 6 months.
Accession Number P21583.1
Name ProDots™ Recombinant Human SCF