Please login to your online account to display your discounted pricing

R&D Systems ProDots Recombinant Human FGF basic (146 aa)


Pre-aliquoted balls of FGF basic protein for cell culture media

Manufacturer: r&d systems  PRD23350

 View more versions of this product

Catalog No. PRD23350

  • / Each
 

Description & Specifications

Specifications

Quantity 250μg
Purity >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight 16.5kDa
Storage Requirements Stored at 4°C where they will remain stable for 6 months.
Recombinant Yes
Source E. coli-derived, Pro143-Ser288, with an N-terminal Ala
Name ProDots™ Recombinant Human FGF basic
Accession Number P09038