missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Porcine TNF-alpha Recombinant Protein

$481.01 - $481.01

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 397086
Molecular Weight 17 kDa
Name TNF-alpha
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
690PT025CF R&D Systems
RDS690PT025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $481.01
690PT025 R&D Systems
RDS690PT025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $481.01
Specifications

Description

Source: E. coli-derived, Arg78-Leu232, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 397086
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
17 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
397086
TNF-alpha
25μg
Porcine