Please login to your online account to display your discounted pricing

R&D Systems Porcine TGF-beta 2 Protein

$481.01 - $481.01

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 397084
Name TGF-beta 2
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 1μg
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

102B2001

 
r&d systems
RDS102B2001
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $481.01

102B2001CF

 
r&d systems
RDS102B2001CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $481.01
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 397084
Name TGF-beta 2
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 1μg
Source Porcine

Source: Porcine platelet-derived
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 397084
Regulatory Status: RUO