R&D Systems Porcine TGF-beta 2 Protein
Manufacturer:  R&D Systems RDS102B2001CF
 View more versions of this product
Catalog No. 102B2001CF
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
TGF-beta 2
1μg
Disulfide-linked homodimer
397084
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Porcine
Unlabeled