missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Porcine TGF-beta 1 Protein

$337.62 - $2,071.36

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 397078
Name TGF-beta 1
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Source Porcine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
101B1001 R&D Systems
RDS101B1001
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 1μg Each for $337.62
101B1001CF R&D Systems
RDS101B1001CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 1μg Each for $337.62
101B1010 R&D Systems
RDS101B1010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 10μg Each for $2,071.36
101B1010CF R&D Systems
RDS101B1010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 10μg Each for $2,071.36
Specifications

Description

Source: Porcine platelet-derived
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 397078
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
TGF-beta 1
Porcine
397078
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.