Please login to your online account to display your discounted pricing

R&D Systems Porcine TGF-beta 1 Protein


Manufacturer: r&d systems  RDS101B1010

 View more versions of this product

Catalog No. 101B1010

  • / Each
 

Description & Specifications

Specifications

Quantity 10μg
Name TGF-beta 1
Gene ID (Entrez) 397078
Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Structural Form Disulfide-linked homodimer
Source Porcine
Conjugate Unlabeled
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.