Please login to your online account to display your discounted pricing

R&D Systems Porcine TGF-beta 1 Protein


Manufacturer: r&d systems  RDS101B1001CF

 View more versions of this product

Catalog No. 101B1001CF

  • / Each
 

Description & Specifications

Specifications

Structural Form Disulfide-linked homodimer
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Gene ID (Entrez) 397078
Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Name TGF-beta 1
Source Porcine
Conjugate Unlabeled
Quantity 1μg