missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Porcine IL-6 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 399500
Molecular Weight 21 kDa
Name IL-6
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
686PI025 R&D Systems
RDS686PI025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $428.87
686PI025CF R&D Systems
RDS686PI025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $428.87
Specifications

Description

Source: E. coli-derived, Pro29-Met212
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 21 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 399500
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
21 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
399500
IL-6
25μg
Porcine