missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Porcine IL-5 Recombinant Protein

$389.76 - $389.76

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 397409
Molecular Weight 14 kDa (monomer)
Name IL-5
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
3137PL010 R&D Systems
RDS3137PL010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $389.76
3137PL010CF R&D Systems
RDS3137PL010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $389.76
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ile20-Ser134, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 397409
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
14 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
397409
IL-5
10μg
Porcine