missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Porcine IL-4 Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 397225
Molecular Weight 12.5 kDa
Name IL-4
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
654P4025CF R&D Systems
RDS654P4025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $416.79
654P4025 R&D Systems
RDS654P4025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $416.79
Specifications

Description

Source: E. coli-derived, His25-Cys133, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 397225
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
12.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
397225
IL-4
25μg
Porcine