missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Porcine IL-1ra/IL-1F3 Recombinant Protein

$454.80 - $454.80

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 397499
Molecular Weight 17 kDa
Name IL-1ra/IL-1F3
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
780RA025 R&D Systems
RDS780RA025
Lyophilized from a 0.2μm filtered solution in PBS and DTT with BSA as a carrier protein. Each for $454.80
780RA025CF R&D Systems
RDS780RA025CF
Lyophilized from a 0.2μm filtered solution in PBS and DTT. Each for $454.80
Specifications

Description

Source: E. coli-derived, His26-Gln177, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and DTT.
Entrez Gene ID: 397499
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
17 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
397499
IL-1ra/IL-1F3
25μg
Porcine