Please login to your online account to display your discounted pricing

R&D Systems Porcine IL-18/IL-1F4 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 397057
Molecular Weight 18.0 kDa
Name IL-18/IL-1F4
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

588PL025CF

 
r&d systems
RDS588PL025CF
Lyophilized from a 0.2μm filtered solution in PBS and DTT with Trehalose. Each for $428.87

588PL025

 
r&d systems
RDS588PL025
Lyophilized from a 0.2μm filtered solution in PBS and DTT with Trehalose and with BSA as a carrier protein. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 397057
Molecular Weight 18.0 kDa
Name IL-18/IL-1F4
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Porcine

Source: E. coli-derived, Tyr36-Asn192
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.0 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and DTT with Trehalose.
Entrez Gene ID: 397057
Regulatory Status: RUO