missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Porcine IL-10 Recombinant Protein

$389.76 - $389.76

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 397106
Molecular Weight 18 kDa
Name IL-10
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
693PI010 R&D Systems
RDS693PI010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $389.76
693PI010CF R&D Systems
RDS693PI010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $389.76
Specifications

Description

Source: E. coli-derived, Ser19-Asn175
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 397106
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
397106
IL-10
10μg
Porcine