missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Porcine IL-1 alpha/IL-1F1 Recombinant Protein

$337.62 - $337.62

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 397094
Molecular Weight 17.5 kDa
Name IL-1 alpha/IL-1F1
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
680PI010 R&D Systems
RDS680PI010
Lyophilized from a 0.2μm filtered solution in PBS and DTT with BSA as a carrier protein. Each for $337.62
680PI010CF R&D Systems
RDS680PI010CF
Lyophilized from a 0.2μm filtered solution in PBS and DTT. Each for $337.62
Specifications

Description

Source: E. coli-derived, Gln119-Ser270 with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and DTT.
Entrez Gene ID: 397094
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
17.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
397094
IL-1 alpha/IL-1F1
10μg
Porcine