missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Wnt-9b Recombinant Protein, Ala24-Arg359

$481.01 - $481.01

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 22412
Molecular Weight 36.8 kDa
Name Wnt-9b
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
3669WN025 R&D Systems
RDS3669WN025
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein. Each for $481.01
3669WN025CF R&D Systems
RDS3669WN025CF
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS. Each for $481.01
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Ala24-Arg359
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 36.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS.
Entrez Gene ID: 22412
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
36.8 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
22412
Wnt-9b
25μg
Mouse