missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Wnt-9a Recombinant Protein, Tyr30-Gly365

$481.01 - $481.01

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 216795
Molecular Weight 37 kDa
Name Wnt-9a
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
8148WN025CF R&D Systems
RDS8148WN025CF
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS. Each for $481.01
8148WN025 R&D Systems
RDS8148WN025
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein. Each for $481.01
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Tyr30-Gly365
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 37 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS.
Entrez Gene ID: 216795
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
37 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
216795
Wnt-9a
25μg
Mouse