missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Wnt-4 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 22417
Molecular Weight 36.6 kDa
Name Wnt-4
Purity 60%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
475WN005 R&D Systems
RDS475WN005
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein. Each for $428.87
475WN005CF R&D Systems
RDS475WN005CF
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS. Each for $428.87
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Ser23-Arg351
Purity: 60%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 36.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS.
Entrez Gene ID: 22417
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
36.6 kDa
60%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
22417
Wnt-4
5μg
Mouse