Please login to your online account to display your discounted pricing

R&D Systems Mouse Wnt-3a (High Purity) Recombinant Protein

$1,121.62 - $1,121.62

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 22416
Molecular Weight 37.4 kDa
Name Wnt-3a (High Purity)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1324WNP010

 
r&d systems
RDS1324WNP010
Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA and CHAPS with BSA as a carrier protein. Each for $1,121.62

1324WNP010C

 
r&d systems
RDS1324WNP010CF
Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA and CHAPS. Each for $1,121.62
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 22416
Molecular Weight 37.4 kDa
Name Wnt-3a (High Purity)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
Recombinant Yes
Source Mouse

Source: Chinese Hamster Ovary cell line, CHO-derived, Ser19-Lys352
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 37.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA and CHAPS.
Entrez Gene ID: 22416
Regulatory Status: RUO