missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse VEGF R2/KDR/Flk-1 Fc Chimera Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16542
Molecular Weight 110 kDa (monomer)
Name VEGF R2/KDR/Flk-1 Fc Chimera
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
443KD050CF R&D Systems
RDS443KD050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $416.79
443KD050 R&D Systems
RDS443KD050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $416.79
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ala20-Glu726
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 110 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 16542
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
110 kDa (monomer)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
16542
VEGF R2/KDR/Flk-1 Fc Chimera
50μg
Mouse