missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse VEGF R1/Flt-1 Fc Chimera Recombinant Protein

$416.79 - $454.80

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 14254
Name VEGF R1/Flt-1 Fc Chimera
Recombinant Yes
Source Mouse
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Molecular Weight Purity Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Molecular Weight Purity Quantity Price Quantity  
471F1100 R&D Systems
RDS471F1100
Lyophilized from a 0.2μm filtered solution in MOPS, NaCl and CHAPS. 110 kDa (monomer) 90%, by SDS-PAGE under reducing conditions and visualized by silver stain. 100μg Each for $454.80
7756FL050 R&D Systems
RDS7756FL050
Lyophilized from a 0.2μm filtered solution in PBS. 109.6 kDa (monomer) 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. 50μg Each for $416.79
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ser27-Glu759
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 109.6 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 14254
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
VEGF R1/Flt-1 Fc Chimera
Mouse
14254
Yes