Please login to your online account to display your discounted pricing

R&D Systems Mouse VEGF-D Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 14205
Molecular Weight 13 kDa
Name VEGF-D
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

469VD025

 
r&d systems
RDS469VD025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87

469VD025CF

 
r&d systems
RDS469VD025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 14205
Molecular Weight 13 kDa
Name VEGF-D
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Mouse

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Phe98-Ser206, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 13 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 14205
Regulatory Status: RUO