missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse VEGF-B 186 Recombinant Protein

$389.76 - $389.76

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 22339
Molecular Weight 15.2 kDa
Name VEGF-B 186
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
767VE010 R&D Systems
RDS767VE010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $389.76
767VE010CF R&D Systems
RDS767VE010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $389.76
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (stably transfected)-derived, Gln20-Ala207 & Gln20-Arg148
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.2 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 22339
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
15.2 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
22339
VEGF-B 186
10μg
Mouse