missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse VEGF 188 Recombinant Protein

$376.73 - $376.73

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 22339
Molecular Weight 22.1 kDa (monomer)
Name VEGF 188
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
7916MV010CF R&D Systems
RDS7916MV010CF
Lyophilized from a 0.2μm filtered solution in HCl. Each for $376.73
7916MV010 R&D Systems
RDS7916MV010
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. Each for $376.73
Specifications

Description

Source: Human embryonic kidney cell, HEK293-derived, Ala27-Arg214
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 22.1 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in HCl.
Entrez Gene ID: 22339
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
22.1 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
22339
VEGF 188
10μg
Mouse