missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse VEGF 164 Recombinant Protein

$337.62 - $1,080.65

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 22339
Molecular Weight 19.4 kDa (monomer)
Name VEGF 164
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
493MV025CF R&D Systems
RDS493MV025CF
Lyophilized from a 0.2μm filtered solution in PBS. 25μg Each for $1,080.65
493MV005 R&D Systems
RDS493MV005
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 5μg Each for $337.62
493MV025 R&D Systems
RDS493MV025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 25μg Each for $1,080.65
493MV005CF R&D Systems
RDS493MV005CF
Lyophilized from a 0.2μm filtered solution in PBS. 5μg Each for $337.62
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Ala27-Arg190
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19.4 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 22339
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
19.4 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
22339
VEGF 164
Yes