missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse VEGF 120 Recombinant Protein

$328.11 - $1,050.21

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 22339
Name VEGF 120
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Molecular Weight Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Molecular Weight Quantity Price Quantity  
494VE005 R&D Systems
RDS494VE005
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 14 kDa 5μg Each for $328.11
494VE005CF R&D Systems
RDS494VE005CF
Lyophilized from a 0.2μm filtered solution in PBS. 14 kDa (monomer) 5μg Each for $328.11
494VE025 R&D Systems
RDS494VE025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 14 kDa 25μg Each for $1,050.21
494VE025CF R&D Systems
RDS494VE025CF
Lyophilized from a 0.2μm filtered solution in PBS. 14 kDa (monomer) 25μg Each for $1,050.21
Specifications

Description

Source: E. coli-derived, Ala27-Arg146 (Gly117Glu & Glu118Arg)
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 22339
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
VEGF 120
Yes
22339
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse