missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse VCAM-1/CD106 Fc Chimera Recombinant Protein

$298.51 - $885.12

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 22329
Molecular Weight 101.9 kDa (monomer)
Name VCAM-1/CD106 Fc Chimera
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
643VM050 R&D Systems
RDS643VM050
50μg Each for $298.51
643VM200 R&D Systems
RDS643VM200
200μg Each for $885.12
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Phe25-Glu698
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 101.9 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 22329
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
22329
VCAM-1/CD106 Fc Chimera
Yes
Lyophilized from a 0.2μm filtered solution in PBS.
101.9 kDa (monomer)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse