Please login to your online account to display your discounted pricing

R&D Systems Mouse ULBP-1 /MULT-1 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 77777
Molecular Weight 22 kDa (monomer)
Name ULBP-1 /MULT-1
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

2588MU050

 
r&d systems
RDS2588MU050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87

2588MU050CF

 
r&d systems
RDS2588MU050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 77777
Molecular Weight 22 kDa (monomer)
Name ULBP-1 /MULT-1
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
Recombinant Yes
Source Mouse

Source: Mouse myeloma cell line, NS0-derived, Pro26-Thr211, with a C-terminal 6-His tag
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 22 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 77777
Regulatory Status: RUO