missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse TWEAK/TNFSF12 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 21944
Molecular Weight 17 kDa
Name TWEAK/TNFSF12
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1237TW025 R&D Systems
RDS1237TW025
Lyophilized from a 0.2μm filtered solution in PBS and NaCl with BSA as a carrier protein. Each for $428.87
1237TW025CF R&D Systems
RDS1237TW025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: E. coli-derived, Arg105-His249, with an N-terminal Met and a 6-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 21944
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
17 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
21944
TWEAK/TNFSF12
25μg
Mouse