Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
missing translation for 'pleaseLogIn'

R&D Systems Mouse TrkC Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 18213
Molecular Weight 46 kDa
Name TrkC
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Qty  
Catalog Number Mfr. No. Formulation Price Qty  
1404TC050 R&D Systems
RDS1404TC050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
1404TC050CF R&D Systems
RDS1404TC050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Cys32-Thr429, with a C-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 46 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 18213
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
46 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
18213
TrkC
50μg
Mouse